Product Details

SNP ID
rs138927383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111702264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGATTTATGAGAGCAGAAATAA[C/T]AGCAGAGGGTTTTTTACGAGAATTT
Phenotype
MIM: 611181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAD10 PubMed Links

Gene Details

Gene
ACAD10
Gene Name
acyl-CoA dehydrogenase family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136538.1 490 Missense Mutation ACA,ATA T97I NP_001130010.1
NM_025247.5 490 Missense Mutation ACA,ATA T97I NP_079523.3

View Full Product Details