Product Details

SNP ID
rs139088741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120343015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCAGGCAGTAGCGGGTCCGAGT[C/T]GCTGCAGGGGAGGGGGCGGCGGCTG
Phenotype
MIM: 603328
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSI1 PubMed Links

Gene Details

Gene
MSI1
Gene Name
musashi RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002442.3 1961 UTR 3 NP_002433.1
XM_006719403.3 1961 UTR 3 XP_006719466.1
XM_006719404.3 1961 UTR 3 XP_006719467.1
XM_011538361.2 1961 Intron XP_011536663.1
XM_011538362.2 1961 UTR 3 XP_011536664.1
XM_011538363.2 1961 UTR 3 XP_011536665.1
XM_011538364.2 1961 UTR 3 XP_011536666.1
XM_011538365.2 1961 UTR 3 XP_011536667.1
XM_011538366.2 1961 UTR 3 XP_011536668.1
XM_011538368.2 1961 UTR 3 XP_011536670.1
XM_011538370.2 1961 UTR 3 XP_011536672.1
XM_011538371.2 1961 UTR 3 XP_011536673.1

View Full Product Details