Product Details

SNP ID
rs139160089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14942648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCAGCATGCCCTTGGGAGCCTC[C/T]TCAACTGGAGTGAGGAACTCATACT
Phenotype
MIM: 602843 MIM: 610642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGDIB PubMed Links

Gene Details

Gene
ARHGDIB
Gene Name
Rho GDP dissociation inhibitor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001175.6 593 Silent Mutation GAA,GAG E160E NP_001166.3
NM_001321420.1 593 Silent Mutation GAA,GAG E160E NP_001308349.1
NM_001321421.1 593 Silent Mutation GAA,GAG E160E NP_001308350.1
NM_001321422.1 593 Silent Mutation GAA,GAG E160E NP_001308351.1
NM_001321423.1 593 Silent Mutation GAA,GAG E160E NP_001308352.1
Gene
ERP27
Gene Name
endoplasmic reticulum protein 27
There are no transcripts associated with this gene.

View Full Product Details