Product Details

SNP ID
rs139257369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76861452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTCACTGACTTACTTTTGGGGT[C/T]ATGGATAAATCTGGTTTGGATTGAG
Phenotype
MIM: 601871 MIM: 607799
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSRP2 PubMed Links

Gene Details

Gene
CSRP2
Gene Name
cysteine and glycine rich protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300965.1 Intron NP_001287894.1
NM_001321.2 Intron NP_001312.1
Gene
ZDHHC17
Gene Name
zinc finger DHHC-type containing 17
There are no transcripts associated with this gene.

View Full Product Details