Product Details

SNP ID
rs139264803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95474211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTGGAGGAGGTAGCGGCCTCCG[A/G]GAGCCACCTGAATGGCGACCTGGAT
Phenotype
MIM: 601870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METAP2 PubMed Links

Gene Details

Gene
METAP2
Gene Name
methionyl aminopeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317182.1 166 Missense Mutation GAG,GGG E11G NP_001304111.1
NM_001317183.1 166 Missense Mutation GAG,GGG E11G NP_001304112.1
NM_006838.3 166 Missense Mutation GAG,GGG E11G NP_006829.1
XM_005268583.3 166 Missense Mutation GAG,GGG E11G XP_005268640.1

View Full Product Details