Product Details

SNP ID
rs139289275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104987217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGAAGGAAGTATAATAATGATA[A/G]CCAAAGCTTATGTAGTCCTTACTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf45 PubMed Links

Gene Details

Gene
C12orf45
Gene Name
chromosome 12 open reading frame 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152318.2 Intron NP_689531.2

View Full Product Details