Product Details

SNP ID
rs139363111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:46362353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATATAGAAGGCAGAAGGAAGAAT[A/G]AAAATCAACATAGAAGCTGCAGATG
Phenotype
MIM: 605180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC38A2 PubMed Links

Gene Details

Gene
SLC38A2
Gene Name
solute carrier family 38 member 2
There are no transcripts associated with this gene.

View Full Product Details