Product Details

SNP ID
rs139371126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49003162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTACCTCTGCTTCCACTAGCCT[A/G]TTGATGATGGTCTCCAGAGTCTCAT
Phenotype
MIM: 610588 MIM: 602742
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDN PubMed Links

Gene Details

Gene
DDN
Gene Name
dendrin
There are no transcripts associated with this gene.

Gene
PRKAG1
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206709.1 976 Silent Mutation AAC,AAT N299N NP_001193638.1
NM_001206710.1 976 Silent Mutation AAC,AAT N258N NP_001193639.1
NM_002733.4 976 Silent Mutation AAC,AAT N290N NP_002724.1
XM_005269019.4 976 Silent Mutation AAC,AAT N327N XP_005269076.1
XM_005269020.2 976 Silent Mutation AAC,AAT N206N XP_005269077.1
XM_006719499.2 976 Silent Mutation AAC,AAT N258N XP_006719562.1
XM_011538562.2 976 Silent Mutation AAC,AAT N258N XP_011536864.1
XM_017019622.1 976 Silent Mutation AAC,AAT N206N XP_016875111.1
XM_017019623.1 976 Silent Mutation AAC,AAT N206N XP_016875112.1

View Full Product Details