Product Details

SNP ID
rs139471286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:67655128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATGTATTAAATTATGCGAAACA[C/T]AAAAATTGCTTGAAGTAAATTTCTG
Phenotype
MIM: 603496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYRK2 PubMed Links

Gene Details

Gene
DYRK2
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003583.3 Intron NP_003574.1
NM_006482.2 Intron NP_006473.2
XM_017020032.1 Intron XP_016875521.1

View Full Product Details