Product Details

SNP ID
rs139738994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121652767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCTGTCTTCCTGAACATGGC[C/T]AAGGCCTCCGCCAGCACACCATCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MORN3 PubMed Links

Gene Details

Gene
MORN3
Gene Name
MORN repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173855.4 801 Silent Mutation TTA,TTG L230L NP_776254.3
XM_005253869.4 801 Silent Mutation TTA,TTG L230L XP_005253926.1
XM_011538213.2 801 Silent Mutation TTA,TTG L230L XP_011536515.1
XM_011538214.2 801 Silent Mutation TTA,TTG L115L XP_011536516.1

View Full Product Details