Product Details

SNP ID
rs139893949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49951040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCGGGGCCCACATCAACCCTGC[C/T]GTGACTGTGGCCTGCCTGGTGGGCT
Phenotype
MIM: 107777
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP2 PubMed Links

Gene Details

Gene
AQP2
Gene Name
aquaporin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000486.5 300 Silent Mutation GCC,GCT A70A NP_000477.1
Gene
LOC101927318
Gene Name
uncharacterized LOC101927318
There are no transcripts associated with this gene.

View Full Product Details