Product Details

SNP ID
rs139954107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52645219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCTTAGAGCCACCACCAGATC[C/T]GTATCTTCCTCCAGAACCACCGCCA
Phenotype
MIM: 600194
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT2 PubMed Links

Gene Details

Gene
KRT2
Gene Name
keratin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000423.2 1753 Missense Mutation AGA,GGA R574G NP_000414.2

View Full Product Details