Product Details

SNP ID
rs139960283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:11185997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCTTCTGTAAAGTCTGTCTAC[A/C]AAGGTAAAGGGTTTGGTGTTTTTGT
Phenotype
MIM: 613966
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMIM10L1 PubMed Links

Gene Details

Gene
SMIM10L1
Gene Name
small integral membrane protein 10 like 1
There are no transcripts associated with this gene.

Gene
TAS2R42
Gene Name
taste 2 receptor member 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181429.2 941 Missense Mutation TGG,TTG W314L NP_852094.2

View Full Product Details