Product Details

SNP ID
rs140167393
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12718116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGAGTTCTACTACAGACCCCCG[C/T]GGCCCCCCAAAGGTGCCTGCAAGGT
Phenotype
MIM: 612456 MIM: 600778 MIM: 602927
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
APOLD1 PubMed Links

Gene Details

Gene
APOLD1
Gene Name
apolipoprotein L domain containing 1
There are no transcripts associated with this gene.

Gene
CDKN1B
Gene Name
cyclin dependent kinase inhibitor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004064.4 847 Missense Mutation CGG,TGG R93W NP_004055.1
Gene
GPR19
Gene Name
G protein-coupled receptor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006143.2 847 Intron NP_006134.1

View Full Product Details