Product Details

SNP ID
rs140263819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:70521449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAGTTTTTAAAAACAAATCACA[C/G]AGTGAATAATTTTTATCCAGGAGCT
Phenotype
MIM: 176882
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTPRB PubMed Links

Gene Details

Gene
PTPRB
Gene Name
protein tyrosine phosphatase, receptor type B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006719528.1 6469 Intron XP_006719591.1
XM_006719529.3 6469 Intron XP_006719592.1
XM_011538614.1 6469 Intron XP_011536916.1
XM_017019723.1 6469 UTR 3 XP_016875212.1
XM_017019724.1 6469 Intron XP_016875213.1

View Full Product Details