Product Details

SNP ID
rs140277271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6945853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTGTCAGGCAAGCTGAAGGC[A/G]CTGTTTCTGCCGCCCATGACCCTGC
Phenotype
MIM: 607462 MIM: 615140 MIM: 176883
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATN1 PubMed Links

Gene Details

Gene
ATN1
Gene Name
atrophin 1
There are no transcripts associated with this gene.

Gene
C12orf57
Gene Name
chromosome 12 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301834.1 617 Silent Mutation GCA,GCG A104A NP_001288763.1
NM_001301836.1 617 Silent Mutation GCA,GCG A91A NP_001288765.1
NM_001301837.1 617 Silent Mutation GCA,GCG A75A NP_001288766.1
NM_001301838.1 617 Silent Mutation GCA,GCG A69A NP_001288767.1
NM_138425.3 617 Silent Mutation GCA,GCG A104A NP_612434.1
Gene
PTPN6
Gene Name
protein tyrosine phosphatase, non-receptor type 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002831.5 617 Intron NP_002822.2
NM_080548.4 617 Intron NP_536858.1
NM_080549.3 617 Intron NP_536859.1
XM_006718994.1 617 Intron XP_006719057.1
XM_011520988.1 617 Intron XP_011519290.1
Gene
RNU7-1
Gene Name
RNA, U7 small nuclear 1
There are no transcripts associated with this gene.

View Full Product Details