Product Details
- SNP ID
-
rs140358670
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6668020 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCCTGGGCTTGAGCTTGAGCCTG[A/G]GCCTGGGCCACTGCTGCCGCTGCTG
- Phenotype
-
MIM: 608524
MIM: 609951
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ING4
PubMed Links
Gene Details
- Gene
- ING4
- Gene Name
- inhibitor of growth family member 4
There are no transcripts associated with this gene.
- Gene
- ZNF384
- Gene Name
- zinc finger protein 384
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039920.2 |
1768 |
Silent Mutation |
GCC,GCT |
A360A |
NP_001035009.1 |
NM_001135734.2 |
1768 |
Silent Mutation |
GCC,GCT |
A476A |
NP_001129206.1 |
NM_133476.4 |
1768 |
Silent Mutation |
GCC,GCT |
A415A |
NP_597733.2 |
XM_006718969.3 |
1768 |
Silent Mutation |
GCC,GCT |
A245A |
XP_006719032.1 |
XM_011520917.1 |
1768 |
Silent Mutation |
GCC,GCT |
A507A |
XP_011519219.1 |
XM_011520918.1 |
1768 |
Silent Mutation |
GCC,GCT |
A507A |
XP_011519220.1 |
XM_011520919.1 |
1768 |
Silent Mutation |
GCC,GCT |
A507A |
XP_011519221.1 |
XM_011520920.1 |
1768 |
Silent Mutation |
GCC,GCT |
A507A |
XP_011519222.1 |
XM_011520921.2 |
1768 |
Silent Mutation |
GCC,GCT |
A507A |
XP_011519223.1 |
XM_011520922.1 |
1768 |
Silent Mutation |
GCC,GCT |
A491A |
XP_011519224.1 |
XM_011520923.1 |
1768 |
Silent Mutation |
GCC,GCT |
A491A |
XP_011519225.1 |
XM_017018941.1 |
1768 |
Silent Mutation |
GCC,GCT |
A476A |
XP_016874430.1 |
XM_017018942.1 |
1768 |
Silent Mutation |
GCC,GCT |
A476A |
XP_016874431.1 |
XM_017018943.1 |
1768 |
Silent Mutation |
GCC,GCT |
A476A |
XP_016874432.1 |
XM_017018944.1 |
1768 |
Silent Mutation |
GCC,GCT |
A460A |
XP_016874433.1 |
XM_017018945.1 |
1768 |
Silent Mutation |
GCC,GCT |
A460A |
XP_016874434.1 |
XM_017018946.1 |
1768 |
Silent Mutation |
GCC,GCT |
A446A |
XP_016874435.1 |
XM_017018947.1 |
1768 |
Silent Mutation |
GCC,GCT |
A446A |
XP_016874436.1 |
XM_017018948.1 |
1768 |
Silent Mutation |
GCC,GCT |
A430A |
XP_016874437.1 |
XM_017018949.1 |
1768 |
Silent Mutation |
GCC,GCT |
A415A |
XP_016874438.1 |
XM_017018950.1 |
1768 |
Silent Mutation |
GCC,GCT |
A415A |
XP_016874439.1 |
XM_017018951.1 |
1768 |
Silent Mutation |
GCC,GCT |
A399A |
XP_016874440.1 |
XM_017018952.1 |
1768 |
Silent Mutation |
GCC,GCT |
A399A |
XP_016874441.1 |
XM_017018953.1 |
1768 |
Silent Mutation |
GCC,GCT |
A399A |
XP_016874442.1 |
XM_017018954.1 |
1768 |
Silent Mutation |
GCC,GCT |
A331A |
XP_016874443.1 |
View Full Product Details