Product Details

SNP ID
rs140601580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4720803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACCTGTTTACGGGTGGTCTCCA[C/T]AGGGAGCTTCCTTTACATCTGAATA
Phenotype
MIM: 606250
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNT8 PubMed Links

Gene Details

Gene
GALNT8
Gene Name
polypeptide N-acetylgalactosaminyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017417.1 218 Silent Mutation CAC,CAT H42H NP_059113.1
Gene
LOC101929549
Gene Name
uncharacterized LOC101929549
There are no transcripts associated with this gene.

View Full Product Details