Product Details
- SNP ID
-
rs140654887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:12975339 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCAACAGCCAGATCTCATTGCGC[C/T]GTCCGTAGGGCTTCATGGGAGGGTC
- Phenotype
-
MIM: 605826
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM234B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1941] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM234B
- Gene Name
- family with sequence similarity 234 member B
- Gene
- HEBP1
- Gene Name
- heme binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015987.4 |
734 |
Missense Mutation |
CAG,CGG |
Q180R |
NP_057071.2 |
- Gene
- LOC100506314
- Gene Name
- uncharacterized LOC100506314
There are no transcripts associated with this gene.
View Full Product Details