Product Details

SNP ID
rs140827395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7304392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGGCTCACCAAGCCACCTGGC[C/T]GGCGCTTACACAAAGATCACCAGCT
Phenotype
MIM: 614360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSM4 PubMed Links

Gene Details

Gene
ACSM4
Gene Name
acyl-CoA synthetase medium-chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080454.1 61 Missense Mutation CGG,TGG R21W NP_001073923.1

View Full Product Details