Product Details
- SNP ID
-
rs140843522
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:88495184 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTACTAGGTTTGAGGACAAAAG[C/T]ACACATTTATTATACTCTTAAGCAC
- Phenotype
-
MIM: 184745
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KITLG
PubMed Links
Gene Details
- Gene
- KITLG
- Gene Name
- KIT ligand
View Full Product Details