Product Details

SNP ID
rs140968898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55365385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACAGGAAACAGAACCATTTCTT[A/G]TAATGGGTGTGTGGCTCAGCTATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6C75 PubMed Links

Gene Details

Gene
OR6C75
Gene Name
olfactory receptor family 6 subfamily C member 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005497.1 275 Missense Mutation TAT,TGT Y92C NP_001005497.1

View Full Product Details