Product Details
- SNP ID
-
hCV161850122
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:121652776 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCCTGAACATGGCCAAGGCCTC[A/C]GCCAGCACACCATCAGGGTCTAGGA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MORN3
PubMed Links
Gene Details
- Gene
- MORN3
- Gene Name
- MORN repeat containing 3
There are no transcripts associated with this gene.
View Full Product Details