Product Details

SNP ID
rs141204840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113157516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGATCTCCAGTCCCCGCCCTA[C/G]CTTTCGGCCTCCCCCGCGTGTTGAG
Phenotype
MIM: 611665
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP73 PubMed Links

Gene Details

Gene
CFAP73
Gene Name
cilia and flagella associated protein 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144872.1 4035 Intron NP_001138344.1
XM_011538327.2 4035 Intron XP_011536629.1
Gene
DDX54
Gene Name
DEAD-box helicase 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111322.1 4035 UTR 3 NP_001104792.1
NM_024072.3 4035 UTR 3 NP_076977.3
Gene
MIR7106
Gene Name
microRNA 7106
There are no transcripts associated with this gene.

View Full Product Details