Product Details

SNP ID
rs141212020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130163658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCATCTGGGCGGTGCTGTGCT[G/T]CTTCTCCAGCGCCTTCACCGTGCTC
Phenotype
MIM: 606147
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FZD10 PubMed Links

Gene Details

Gene
FZD10
Gene Name
frizzled class receptor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007197.3 1200 Missense Mutation TGC,TTC C239F NP_009128.1
Gene
FZD10-AS1
Gene Name
FZD10 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details