Product Details

SNP ID
rs141382861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52608006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGATGGAGGCAGAAAGATGGGCT[C/T]CAGCTCTCAAATCCTGATTCAACAT
Phenotype
MIM: 608246 MIM: 608247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT72 PubMed Links

Gene Details

Gene
KRT72
Gene Name
keratin 72
There are no transcripts associated with this gene.

Gene
KRT73
Gene Name
keratin 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175068.2 3326 UTR 3 NP_778238.1
XM_011538257.2 3326 UTR 3 XP_011536559.1
Gene
KRT73-AS1
Gene Name
KRT73 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details