Product Details

SNP ID
rs141496776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57773033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCGTAGGCCCTGAGCCTGTGCA[A/G]TTATTTCGAGAGTCAAAATGTGGAT
Phenotype
MIM: 609506 MIM: 604466 MIM: 615258 MIM: 604723
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP27B1 PubMed Links

Gene Details

Gene
CYP27B1
Gene Name
cytochrome P450 family 27 subfamily B member 1
There are no transcripts associated with this gene.

Gene
METTL1
Gene Name
methyltransferase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005371.5 319 Intron NP_005362.3
NM_023033.3 319 Intron NP_075422.3
XM_005268873.2 319 Intron XP_005268930.1
XM_017019305.1 319 Intron XP_016874794.1
Gene
METTL21B
Gene Name
methyltransferase like 21B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015433.2 319 Missense Mutation AAT,AGT N65S NP_056248.2
NM_206914.1 319 Missense Mutation AAT,AGT N65S NP_996797.1
Gene
TSFM
Gene Name
Ts translation elongation factor, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details