Product Details
- SNP ID
-
rs141650005
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:51048366 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCTCGCTGTGAAGATGGCGCTC[A/T]CCAGGGTGTGCTGGGCTCGGTCGGC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LETMD1
PubMed Links
Gene Details
- Gene
- LETMD1
- Gene Name
- LETM1 domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001243689.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
NP_001230618.1 |
NM_001300765.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
NP_001287694.1 |
NM_015416.4 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
NP_056231.3 |
XM_006719335.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_006719398.1 |
XM_006719336.1 |
68 |
UTR 5 |
|
|
XP_006719399.1 |
XM_006719339.1 |
68 |
UTR 5 |
|
|
XP_006719402.1 |
XM_006719340.1 |
68 |
UTR 5 |
|
|
XP_006719403.1 |
XM_011538161.2 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_011536463.1 |
XM_011538162.2 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_011536464.1 |
XM_011538163.2 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_011536465.1 |
XM_011538164.2 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_011536466.1 |
XM_011538165.2 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_011536467.1 |
XM_011538167.1 |
68 |
Missense Mutation |
CAC,CTC |
H41L |
XP_011536469.1 |
XM_017019151.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874640.1 |
XM_017019152.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874641.1 |
XM_017019153.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874642.1 |
XM_017019154.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874643.1 |
XM_017019155.1 |
68 |
UTR 5 |
|
|
XP_016874644.1 |
XM_017019156.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874645.1 |
XM_017019157.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874646.1 |
XM_017019158.1 |
68 |
UTR 5 |
|
|
XP_016874647.1 |
XM_017019159.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874648.1 |
XM_017019160.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874649.1 |
XM_017019161.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874650.1 |
XM_017019162.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874651.1 |
XM_017019163.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874652.1 |
XM_017019164.1 |
68 |
UTR 5 |
|
|
XP_016874653.1 |
XM_017019165.1 |
68 |
UTR 5 |
|
|
XP_016874654.1 |
XM_017019166.1 |
68 |
UTR 5 |
|
|
XP_016874655.1 |
XM_017019167.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874656.1 |
XM_017019168.1 |
68 |
Missense Mutation |
ACC,TCC |
T4S |
XP_016874657.1 |
View Full Product Details