Product Details

SNP ID
rs141682565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:75481887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGCATGGGCCAGCAATTGCCA[A/G]TTTTCACATAATACACGGCTGAAGC
Phenotype
MIM: 602692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLIPR1 PubMed Links

Gene Details

Gene
GLIPR1
Gene Name
GLI pathogenesis related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006851.2 355 Silent Mutation CAA,CAG Q76Q NP_006842.2
XM_011537775.2 355 Silent Mutation CAA,CAG Q76Q XP_011536077.1
XM_017018721.1 355 UTR 5 XP_016874210.1

View Full Product Details