Product Details

SNP ID
rs141775673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130398187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGTATTTAAATATACGTTAAGC[A/G]CTTAATAGTGCTTCAGAGGGGTAGA
Phenotype
MIM: 605571 MIM: 611602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIWIL1 PubMed Links

Gene Details

Gene
PIWIL1
Gene Name
piwi like RNA-mediated gene silencing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190971.1 4271 Intron NP_001177900.1
NM_004764.4 4271 Intron NP_004755.2
XM_011539002.2 4271 Intron XP_011537304.1
XM_011539003.2 4271 Intron XP_011537305.1
XM_011539004.2 4271 Intron XP_011537306.1
XM_011539005.1 4271 Intron XP_011537307.1
XM_011539006.2 4271 Intron XP_011537308.1
XM_017020229.1 4271 Intron XP_016875718.1
Gene
RIMBP2
Gene Name
RIMS binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015347.4 4271 UTR 3 NP_056162.4
XM_011538102.1 4271 Intron XP_011536404.1
XM_011538103.2 4271 Intron XP_011536405.1
XM_011538104.2 4271 Intron XP_011536406.1
XM_011538105.2 4271 Intron XP_011536407.1
XM_011538106.2 4271 Intron XP_011536408.1
XM_011538107.2 4271 Intron XP_011536409.1
XM_011538108.2 4271 Intron XP_011536410.1
XM_017019105.1 4271 Intron XP_016874594.1
XM_017019106.1 4271 Intron XP_016874595.1
XM_017019107.1 4271 Intron XP_016874596.1
XM_017019108.1 4271 Intron XP_016874597.1
XM_017019109.1 4271 Intron XP_016874598.1
XM_017019110.1 4271 UTR 3 XP_016874599.1
XM_017019111.1 4271 Intron XP_016874600.1
XM_017019112.1 4271 Intron XP_016874601.1

View Full Product Details