Product Details

SNP ID
rs141908560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57012424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGAGGATGCCAAAGCTGGGGAC[A/G]TTCTCTTGATTCACATCCGTAGGAG
Phenotype
MIM: 162330
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAC3 PubMed Links

Gene Details

Gene
TAC3
Gene Name
tachykinin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178054.1 500 Silent Mutation AAC,AAT N89N NP_001171525.1
NM_013251.3 500 Silent Mutation AAC,AAT N107N NP_037383.1
Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
There are no transcripts associated with this gene.

View Full Product Details