Product Details

SNP ID
rs141911537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69350240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGTAATGATGGCAAAACCCCAG[A/G]AGCAGTTAATGCCTGTCATTTATCC
Phenotype
MIM: 153450 MIM: 602116
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYZ PubMed Links

Gene Details

Gene
LYZ
Gene Name
lysozyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000239.2 324 Missense Mutation GAA,GGA E90G NP_000230.1
Gene
YEATS4
Gene Name
YEATS domain containing 4
There are no transcripts associated with this gene.

View Full Product Details