Product Details

SNP ID
rs141942720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113186746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTCACCAGGGACCACAGGCAG[C/T]ATGAAGACCCCCGTGGAGCTGGCCG
Phenotype
MIM: 611665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX54 PubMed Links

Gene Details

Gene
DDX54
Gene Name
DEAD-box helicase 54
There are no transcripts associated with this gene.

Gene
IQCD
Gene Name
IQ motif containing D
There are no transcripts associated with this gene.

Gene
RITA1
Gene Name
RBPJ interacting and tubulin associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286215.1 664 Silent Mutation AGC,AGT S24S NP_001273144.1
NM_032848.2 664 UTR 5 NP_116237.1
XM_005253972.2 664 UTR 5 XP_005254029.1

View Full Product Details