Product Details

SNP ID
rs142098740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:125002145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCGGTTTATGTCACTTTTTGTCT[A/G]TTGTGAATGGATGCTGCTGTGAATA
Phenotype
MIM: 615627
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRI3BP PubMed Links

Gene Details

Gene
BRI3BP
Gene Name
BRI3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080626.5 Intron NP_542193.3
XM_011537940.2 Intron XP_011536242.1

View Full Product Details