Product Details

SNP ID
rs142239260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56154832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGATCGAGTTTAACAAGGACC[A/T]GCTGGAGGGTGAGGAGAAGCTCATC
Phenotype
MIM: 609931 MIM: 609930 MIM: 601734
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC107987175 PubMed Links

Gene Details

Gene
LOC107987175
Gene Name
uncharacterized LOC107987175
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017020294.1 516 Intron XP_016875783.1
Gene
MYL6
Gene Name
myosin light chain 6
There are no transcripts associated with this gene.

Gene
MYL6B
Gene Name
myosin light chain 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199629.1 516 Missense Mutation CAG,CTG Q65L NP_001186558.1
NM_002475.4 516 Missense Mutation CAG,CTG Q65L NP_002466.1
Gene
SMARCC2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
There are no transcripts associated with this gene.

View Full Product Details