Product Details

SNP ID
rs142369752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47663514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCAAAAATGGAAATCAACCACC[A/G]TATCTTTTCTTGAGTTCTTCGACAG
Phenotype
MIM: 611477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPAP3 PubMed Links

Gene Details

Gene
RPAP3
Gene Name
RNA polymerase II associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146075.1 2003 Silent Mutation TAC,TAT Y629Y NP_001139547.1
NM_001146076.1 2003 Silent Mutation TAC,TAT Y504Y NP_001139548.1
NM_024604.2 2003 Silent Mutation TAC,TAT Y663Y NP_078880.2
XM_011538741.2 2003 Silent Mutation TAC,TAT Y340Y XP_011537043.1

View Full Product Details