Product Details

SNP ID
rs142483515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54555766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTTTGATTCTTCCATCTTCTC[C/T]TTTGCCTCATCTTTCTCTGGGTTCT
Phenotype
MIM: 171891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE1B PubMed Links

Gene Details

Gene
PDE1B
Gene Name
phosphodiesterase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000924.3 Intron NP_000915.1
NM_001165975.2 Intron NP_001159447.1
NM_001288768.1 Intron NP_001275697.1
NM_001288769.1 Intron NP_001275698.1
NM_001315534.1 Intron NP_001302463.1
NM_001315535.1 Intron NP_001302464.1
XM_011538456.1 Intron XP_011536758.1
XM_017019432.1 Intron XP_016874921.1
XM_017019433.1 Intron XP_016874922.1

View Full Product Details