Product Details

SNP ID
rs142505275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31383010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATATAAGAAACAGACTTCAAAAG[A/C]CATGGATGACTTGGAAATAGGATTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DENND5B PubMed Links

Gene Details

Gene
DENND5B
Gene Name
DENN domain containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308339.1 8717 UTR 3 NP_001295268.1
NM_144973.3 8717 UTR 3 NP_659410.3
XM_005253326.2 8717 UTR 3 XP_005253383.1
XM_005253327.3 8717 UTR 3 XP_005253384.1
XM_005253330.3 8717 UTR 3 XP_005253387.1
XM_006719040.3 8717 UTR 3 XP_006719103.1
XM_011520590.2 8717 UTR 3 XP_011518892.1
XM_017018896.1 8717 UTR 3 XP_016874385.1
XM_017018897.1 8717 UTR 3 XP_016874386.1
XM_017018898.1 8717 UTR 3 XP_016874387.1
XM_017018899.1 8717 UTR 3 XP_016874388.1

View Full Product Details