Product Details
- SNP ID
-
rs142546239
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:71125336 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATAGGTTGATGCATCCACAGATTC[A/G]TTTGTTCCCGATCTGGCAATACAGG
- Phenotype
-
MIM: 600769
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN8
PubMed Links
Gene Details
- Gene
- TSPAN8
- Gene Name
- tetraspanin 8
There are no transcripts associated with this gene.
View Full Product Details