Product Details

SNP ID
rs142571907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53295898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGTGGTACAGACCAAGTATGT[C/G]GAAGCCAAGGACTGTCTGAACGTGC
Phenotype
MIM: 611366 MIM: 604143 MIM: 604899
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C12orf10 PubMed Links

Gene Details

Gene
C12orf10
Gene Name
chromosome 12 open reading frame 10
There are no transcripts associated with this gene.

Gene
ESPL1
Gene Name
extra spindle pole bodies like 1, separase
There are no transcripts associated with this gene.

Gene
PFDN5
Gene Name
prefoldin subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002624.3 249 Silent Mutation GTC,GTG V44V NP_002615.2
NM_145897.2 249 Intron NP_665904.1

View Full Product Details