Product Details

SNP ID
rs142692534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120210507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAATTGAAATGGGAAAAGCCAG[C/G]TTGGAAGTTTACAGAGAAAAAATAA
Phenotype
MIM: 602505 MIM: 180510
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PXN PubMed Links

Gene Details

Gene
PXN
Gene Name
paxillin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080855.2 4406 UTR 3 NP_001074324.1
NM_001243756.1 4406 UTR 3 NP_001230685.1
NM_002859.3 4406 UTR 3 NP_002850.2
NM_025157.4 4406 UTR 3 NP_079433.3
XM_005253917.3 4406 UTR 3 XP_005253974.1
XM_006719532.1 4406 UTR 3 XP_006719595.1
XM_006719534.1 4406 UTR 3 XP_006719597.1
XM_006719535.3 4406 UTR 3 XP_006719598.1
XM_006719536.2 4406 UTR 3 XP_006719599.1
XM_011538622.1 4406 UTR 3 XP_011536924.1
XM_011538623.1 4406 UTR 3 XP_011536925.1
XM_017019726.1 4406 UTR 3 XP_016875215.1
XM_017019727.1 4406 UTR 3 XP_016875216.1
XM_017019728.1 4406 UTR 3 XP_016875217.1
XM_017019729.1 4406 UTR 3 XP_016875218.1
XM_017019730.1 4406 Intron XP_016875219.1
XM_017019731.1 4406 UTR 3 XP_016875220.1
XM_017019732.1 4406 Intron XP_016875221.1
XM_017019733.1 4406 UTR 3 XP_016875222.1
XM_017019734.1 4406 UTR 3 XP_016875223.1
XM_017019735.1 4406 UTR 3 XP_016875224.1
XM_017019736.1 4406 UTR 3 XP_016875225.1
XM_017019737.1 4406 UTR 3 XP_016875226.1
XM_017019738.1 4406 UTR 3 XP_016875227.1
XM_017019739.1 4406 UTR 3 XP_016875228.1
XM_017019740.1 4406 UTR 3 XP_016875229.1
XM_017019741.1 4406 UTR 3 XP_016875230.1
XM_017019742.1 4406 UTR 3 XP_016875231.1
XM_017019743.1 4406 UTR 3 XP_016875232.1
Gene
PXN-AS1
Gene Name
PXN antisense RNA 1
There are no transcripts associated with this gene.

Gene
RPLP0
Gene Name
ribosomal protein lateral stalk subunit P0
There are no transcripts associated with this gene.

View Full Product Details