Product Details

SNP ID
rs142767765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110720713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAGCACCCTAGGGCTCTCTTT[A/C]TGTCCTAAAGAAAATGAGCATTTAC
Phenotype
MIM: 176914
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPP1CC PubMed Links

Gene Details

Gene
PPP1CC
Gene Name
protein phosphatase 1 catalytic subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244974.1 1606 Intron NP_001231903.1
NM_002710.3 1606 UTR 3 NP_002701.1
XM_011538504.2 1606 Intron XP_011536806.1
XM_011538505.2 1606 Intron XP_011536807.1

View Full Product Details