Product Details

SNP ID
rs142788295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57455748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAACTCATCCTCCACCCCAGGCAG[C/T]GCTGACCAGAGCCCTCCGGAGACTA
Phenotype
MIM: 165220 MIM: 601233 MIM: 612031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLI1 PubMed Links

Gene Details

Gene
GLI1
Gene Name
GLI family zinc finger 1
There are no transcripts associated with this gene.

Gene
INHBC
Gene Name
inhibin beta C subunit
There are no transcripts associated with this gene.

Gene
INHBE
Gene Name
inhibin beta E subunit
There are no transcripts associated with this gene.

View Full Product Details