Product Details

SNP ID
rs142796616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112033236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGATTGCCTACAATCTCTTACCGG[G/T]GAGAGAGAAGTGTCTTCTAAAATAA
Phenotype
MIM: 612755
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR3657 PubMed Links

Gene Details

Gene
MIR3657
Gene Name
microRNA 3657
There are no transcripts associated with this gene.

Gene
NAA25
Gene Name
N(alpha)-acetyltransferase 25, NatB auxiliary subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024953.3 3511 Silent Mutation TCA,TCC S931S NP_079229.2
XM_005253938.1 3511 Silent Mutation TCA,TCC S795S XP_005253995.1
XM_006719606.2 3511 Silent Mutation TCA,TCC S903S XP_006719669.1
XM_017019977.1 3511 Silent Mutation TCA,TCC S580S XP_016875466.1

View Full Product Details