Product Details

SNP ID
rs142903372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55716116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGGAGGGGGCCCGGCGTACCCA[G/T]CCCCCAGCCCGACGTGACCATGCTG
Phenotype
MIM: 601444 MIM: 155740 MIM: 600536 MIM: 601617
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BLOC1S1 PubMed Links

Gene Details

Gene
BLOC1S1
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001487.3 83 Missense Mutation AGC,ATC S22I NP_001478.2
Gene
BLOC1S1-RDH5
Gene Name
BLOC1S1-RDH5 readthrough
There are no transcripts associated with this gene.

Gene
CD63
Gene Name
CD63 molecule
There are no transcripts associated with this gene.

Gene
ITGA7
Gene Name
integrin subunit alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144996.1 83 Intron NP_001138468.1
NM_001144997.1 83 Intron NP_001138469.1
NM_002206.2 83 Intron NP_002197.2
XM_005268839.1 83 Intron XP_005268896.1
XM_005268840.1 83 Intron XP_005268897.1
XM_005268841.1 83 Intron XP_005268898.1
XM_005268842.1 83 Intron XP_005268899.1
XM_005268844.1 83 Intron XP_005268901.1
XM_005268846.1 83 Intron XP_005268903.1
XM_005268848.1 83 Intron XP_005268905.1
XM_005268849.1 83 Intron XP_005268906.1
XM_005268850.1 83 Intron XP_005268907.1
XM_017019265.1 83 Intron XP_016874754.1
Gene
RDH5
Gene Name
retinol dehydrogenase 5
There are no transcripts associated with this gene.

View Full Product Details