Product Details

SNP ID
rs142985043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50104666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATGTGGGTATTTGAGGAAGACA[C/T]TGGAGGCAAAAAGCTGACTGAGATC
Phenotype
MIM: 614478 MIM: 138420 MIM: 601735
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX14 PubMed Links

Gene Details

Gene
COX14
Gene Name
COX14, cytochrome c oxidase assembly factor
There are no transcripts associated with this gene.

Gene
GPD1
Gene Name
glycerol-3-phosphate dehydrogenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257199.1 366 Missense Mutation ACT,ATT T45I NP_001244128.1
NM_005276.3 366 Missense Mutation ACT,ATT T45I NP_005267.2
Gene
SMARCD1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
There are no transcripts associated with this gene.

View Full Product Details