Product Details

SNP ID
rs143023126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112406819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGCTGGCTCGCAGTTTTCTCAC[A/G]TGCTGACTGAAGGGTTTTTTGCCGT
Phenotype
MIM: 603703
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL6 PubMed Links

Gene Details

Gene
RPL6
Gene Name
ribosomal protein L6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000970.4 687 Intron NP_000961.2
NM_001024662.2 687 Intron NP_001019833.1
NM_001320137.1 687 Intron NP_001307066.1
NM_001320138.1 687 Silent Mutation CAC,CAT H136H NP_001307067.1
NM_001320139.1 687 Intron NP_001307068.1
NM_001320140.1 687 Silent Mutation CAC,CAT H136H NP_001307069.1
NM_001320141.1 687 Silent Mutation CAC,CAT H136H NP_001307070.1
NM_001320142.1 687 Intron NP_001307071.1
XM_017019781.1 687 Silent Mutation CAC,CAT H136H XP_016875270.1
XM_017019782.1 687 Intron XP_016875271.1

View Full Product Details