Product Details
- SNP ID
-
rs143065399
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:71128257 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAATAGTCCCTAACCTTTCTCTAA[C/T]AGCTGCCTGGGAAATTCCACCAGCA
- Phenotype
-
MIM: 600769
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN8
PubMed Links
Gene Details
- Gene
- TSPAN8
- Gene Name
- tetraspanin 8
View Full Product Details