Product Details

SNP ID
rs143295407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12898584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAACCATATATGGCTCAGAGGAA[A/G]CTCTCAAGAGGACTTGGGTCTTTTG
Phenotype
MIM: 604138
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPRC5A PubMed Links

Gene Details

Gene
GPRC5A
Gene Name
G protein-coupled receptor class C group 5 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003979.3 Intron NP_003970.1

View Full Product Details