Product Details
- SNP ID
-
rs143417536
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:55492703 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGCAGACCTCTTTGGGGTAACTG[A/C]ATTTTTTCTTTTGGCTACCATGTCA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR6C68
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7304753] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR6C68
- Gene Name
- olfactory receptor family 6 subfamily C member 68
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005519.2 |
326 |
Missense Mutation |
GAA,GCA |
E109A |
NP_001005519.2 |
View Full Product Details